Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Olfr745 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146299.2 | NP_666411.2 | 1 | 688 | ||
GenBank mRNA | BC132579.1 | - | 1 | 686 | 40130856 | |
BC132581.1 | - | 1 | 686 | 40130858 |
Set Membership: |
![]() |