Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IFNAR2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000874.4 | NP_000865.2 | 6 | 890 | ||
NM_001289125.1 | NP_001276054.1 | 6 | 890 | |||
NM_001289126.1 | NP_001276055.1 | 6 | 844 | |||
NM_001289128.1 | NP_001276057.1 | 6 | 890 | |||
NM_207584.2 | NP_997467.1 | 6 | 844 | |||
NM_207585.2 | NP_997468.1 | 6 | 844 | |||
GenBank mRNA | AK293059.1 | - | 5 | 527 | ||
BC002793.2 | - | 6 | 838 | 3626374 | ||
CR541817.1 | - | 5 | 485 | |||
HG380509.1 | - | 6 | 610 | |||
L41942.1 | - | 6 | 811 | |||
L41943.1 | - | 5 | 524 | |||
L41944.1 | - | 6 | 811 | |||
U29584.1 | - | 6 | 551 | |||
X77722.1 | - | 6 | 710 | |||
X89772.1 | - | 5 | 485 | |||
X89814.1 | - | 6 | 649 |