Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IMPDH2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000884.2 | NP_000875.2 | 5 | 582 | ||
XM_006713128.3 | XP_006713191.1 | 5 | 890 | |||
XM_017006349.1 | XP_016861838.1 | 4 | 767 | |||
XM_017006350.1 | XP_016861839.1 | 4 | 767 | |||
GenBank mRNA | AK293397.1 | - | 543 | |||
AY545560.1 | - | 2 | 200 | |||
BC006124.1 | - | 5 | 530 | 3545693 | ||
BC012840.1 | - | 5 | 523 | 3447994 | ||
BC015567.2 | - | 5 | 521 | 4576285 | ||
J04208.1 | - | 5 | 537 |