Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Olfr13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146652.1 | NP_666863.1 | 1 | 701 | ||
GenBank mRNA | BC120850.1 | - | 1 | 737 | 40129773 |