Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Olfr1443 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146698.2 | NP_666909.1 | 3 | 903 | ||
GenBank mRNA | BC065104.1 | - | 3 | 903 | 6841907 |