Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Olfr902 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146802.2 | NP_667013.2 | 1 | 783 | ||
GenBank mRNA | BC145901.1 | - | 1 | 783 | 40131204 |