Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Olfr895 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146875.2 | NP_667086.2 | 1 | 520 | ||
GenBank mRNA | BC127996.1 | - | 1 | 524 | ||
BC152319.1 | - | 1 | 524 | 40135771 |