Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IKBKE | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001193321.1 | NP_001180250.1 | 21 | 2637 | ||
NM_001193322.1 | NP_001180251.1 | 21 | 2643 | |||
NM_014002.3 | NP_054721.1 | 22 | 2756 | |||
XM_005273356.2 | XP_005273413.1 | 22 | 2752 | |||
XM_017002868.1 | XP_016858357.1 | 21 | 2752 | |||
GenBank mRNA | AK093798.1 | - | 16 | 1617 | ||
BC105923.1 | - | 22 | 2504 | 40001562 | ||
BC105924.1 | - | 22 | 2504 | 40001567 | ||
BC107591.1 | - | 21 | 2391 | |||
BC107812.1 | - | 21 | 2385 | 40001564 | ||
D63485.1 | - | 22 | 2504 |