Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Olfr622 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_147083.1 | NP_667294.1 | 1 | 505 | ||
GenBank mRNA | BC132174.1 | - | 1 | 603 | 40130451 | |
BC132176.1 | - | 1 | 603 | 40130453 |