Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ITPK1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_017021262.1 | XP_016876751.1 | 9 | 2243 | ||
XM_017021263.1 | XP_016876752.1 | 9 | 1969 | |||
XM_017021264.1 | XP_016876753.1 | 8 | 2041 | |||
GenBank mRNA | AF279372.1 | - | 10 | 2107 | ||
AK293515.1 | - | 11 | 1139 | |||
BC018192.1 | - | 11 | 2216 | 3867269 | ||
BC037305.2 | - | 9 | 2133 | |||
U51336.1 | - | 11 | 2136 |