Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PDHB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000925.3 | NP_000916.2 | 3 | 207 | ||
NM_001173468.1 | NP_001166939.1 | 3 | 207 | |||
NM_001315536.1 | NP_001302465.1 | 2 | 153 | |||
NR_033384.1 | 2 | 277 | ||||
GenBank mRNA | AB209241.1 | - | 2 | 1817 | ||
AK225569.1 | - | 3 | 182 | |||
AK225621.1 | - | 3 | 187 | |||
AK293153.1 | - | 3 | 173 | |||
AK311514.1 | - | 3 | 182 | |||
AK313022.1 | - | 3 | 182 | |||
AL117618.1 | - | 2 | 118 | |||
BC000439.2 | - | 3 | 174 | 2820672 | ||
BC001924.1 | - | 3 | 174 | 2820672 | ||
CR541911.1 | - | 3 | 164 | |||
DA965753.1 | - | 3 | 207 | |||
DB039833.1 | - | 2 | 277 | |||
J03576.1 | - | 3 | 182 | |||
M19123.1 | - | 2 | 100 | |||
M34055.1 | - | 3 | 169 | |||
M34056.1 | - | 2 | 92 | |||
M34479.1 | - | 3 | 164 | |||
M54788.1 | - | 3 | 169 | |||
X57778.1 | - | 3 | 183 |