Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc8a2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_148946.2 | NP_683748.1 | 2 | 455 | ||
GenBank mRNA | AF503502.1 | - | 1 | 320 | ||
AK035163.1 | - | 2 | 597 | |||
BC058704.1 | - | 2 | 455 | 6838206 | ||
BC080277.1 | - | 2 | 407 | 30354551 |