Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Ccdc137 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_152807.3 | NP_690020.1 | 3 | 297 | ||
GenBank mRNA | AK156682.1 | - | 3 | 306 | ||
BC026662.1 | - | 3 | 299 | 4989084 |