Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PRRG2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000951.2 | NP_000942.1 | 3 | 401 | ||
NM_001316335.1 | NP_001303264.1 | 3 | 568 | |||
XM_006723286.2 | XP_006723349.1 | 4 | 404 | |||
XM_011527124.1 | XP_011525426.1 | 2 | 396 | |||
GenBank mRNA | AF009243.1 | - | 2 | 245 | ||
BC026032.1 | - | 2 | 595 | 3960215 | ||
CD013954.1 | - | 3 | 548 | |||
CD609900.1 | - | 3 | 409 | |||
CR456846.1 | - | 2 | 236 |