Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gprc5c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001110338.1 | NP_001103808.1 | 4 | 1787 | ||
NM_147217.3 | NP_671750.2 | 4 | 1997 | |||
GenBank mRNA | AK003946.1 | - | 2 | 698 | ||
AK014308.1 | - | 4 | 1793 | |||
AY079516.1 | - | 4 | 1786 | |||
BC031439.1 | - | 4 | 1974 | 4239021 |
Set Membership: |
![]() |