Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Prss44 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_148940.3 | NP_683742.2 | 4 | 803 | ||
GenBank mRNA | AB047758.1 | - | 737 | |||
AB047759.1 | - | 1 | 1230 | |||
AB162857.1 | - | 4 | 789 |