Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Unc5cl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_152823.4 | NP_690036.4 | 9 | 2649 | ||
GenBank mRNA | AK010945.1 | - | 2 | 1310 | ||
AK089034.1 | - | 5 | 1517 | |||
BC022701.1 | - | 5 | 1603 | |||
BC095959.1 | - | 9 | 2627 | 5349225 |