Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Crygn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153076.2 | NP_694716.2 | 2 | 101 | ||
GenBank mRNA | AF445456.1 | - | 2 | 101 | ||
BC120913.1 | - | 2 | 101 | 40091405 | ||
BC120914.1 | - | 1 | 90 | |||
CF739670.1 | - | 1 | 85 |