Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RPS4Y1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001008.3 | NP_000999.1 | 5, 6 | 583 | ||
XM_017030061.1 | XP_016885550.1 | 5, 6 | 667 | |||
GenBank mRNA | AK222594.1 | - | 5, 6 | 564 | ||
AK292819.1 | - | 5, 6 | 568 | |||
BC010286.1 | - | 4, 5 | 552 | 3048929 | ||
BC100905.2 | - | 4, 5 | 540 | 40003602 | ||
BC100906.1 | - | 4, 5 | 538 | |||
M58459.1 | - | 4, 5 | 552 |