Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Cpz | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153107.2 | NP_694747.2 | 10 | 1635 | ||
GenBank mRNA | AF356844.1 | - | 10 | 1544 | ||
BX518878.1 | - | 4 | 462 |