Search Thermo Fisher Scientific
Gene Symbol: Ttc39a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001145948.1 | NP_001139420.1 | 3 | 221 | ||
NM_153392.2 | NP_700441.2 | 3 | 230 | |||
GenBank mRNA | AK048659.1 | - | 3 | 231 | ||
AK076641.1 | - | 3 | 183 | |||
AK076720.1 | - | 3 | 222 | |||
AK076772.1 | - | 3 | 184 | |||
AK082733.1 | - | 3 | 221 | |||
AK172951.1 | - | 3 | 153 | |||
BC023883.1 | - | 3 | 229 | 5150841 | ||
BC026660.1 | - | 3 | 171 | 4988900 | ||
BY095692.1 | - | 3 | 238 |