Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mon2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001163024.1 | NP_001156496.1 | 1, 2 | 662 | ||
NM_001163025.1 | NP_001156497.1 | 1, 2 | 662 | |||
NM_153395.2 | NP_700444.2 | 1, 2 | 662 | |||
GenBank mRNA | AA542162.1 | - | 1, 2 | 150 | ||
AB014059.1 | - | 1, 2 | 184 | |||
AK122425.1 | - | 1, 2 | 662 | |||
AK161217.1 | - | 1, 2 | 541 | |||
BB614144.1 | - | 1, 2 | 541 | |||
BC064131.1 | - | 1, 2 | 152 | |||
BC150943.1 | - | 1, 2 | 183 | 9087854 |