Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Phc3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001165954.1 | NP_001159426.1 | 2, 3 | 213 | ||
NM_001165955.1 | NP_001159427.1 | 2, 3 | 228 | |||
NM_001165956.1 | NP_001159428.1 | 2, 3 | 213 | |||
NM_153421.2 | NP_700470.2 | 2, 3 | 228 | |||
GenBank mRNA | AJ414610.1 | - | 2, 3 | 214 | ||
AK037348.1 | - | 2, 3 | 204 | |||
AK087339.1 | - | 2, 3 | 219 | |||
AK166208.1 | - | 2, 3 | 204 | |||
AK166296.1 | - | 2, 3 | 214 | |||
BC132533.1 | - | 2, 3 | 197 | 40130810 | ||
BC145123.1 | - | 2, 3 | 197 | 9053671 | ||
BC145124.1 | - | 2, 3 | 197 |