Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DAPK2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014326.3 | NP_055141.2 | 3 | 279 | ||
XM_011521413.2 | XP_011519715.1 | 2 | 254 | |||
XM_011521414.1 | XP_011519716.1 | 2 | 254 | |||
XM_011521415.2 | XP_011519717.1 | 2 | 254 | |||
XM_011521416.2 | XP_011519718.1 | 3 | 427 | |||
XM_011521417.1 | XP_011519719.1 | 2 | 254 | |||
XM_017022044.1 | XP_016877533.1 | 2 | 254 | |||
XM_017022045.1 | XP_016877534.1 | 2 | 254 | |||
XM_017022046.1 | XP_016877535.1 | 2 | 254 | |||
GenBank mRNA | AB018001.1 | - | 3 | 278 | ||
AF052941.1 | - | 3 | 279 | |||
AK310967.1 | - | 4 | 366 | |||
BC114506.1 | - | 2 | 268 | 40005390 | ||
GU056176.1 | - | 3 | 279 |