Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Grwd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153419.2 | NP_700468.2 | 4 | 619 | ||
GenBank mRNA | AB001539.1 | - | 5 | 691 | ||
AK077611.1 | - | 4 | 625 | |||
BC008121.1 | - | 4 | 596 | |||
BC028896.1 | - | 3 | 488 | 3674738 | ||
BC083143.1 | - | 4 | 607 |