Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nphp4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153424.2 | NP_700473.2 | 3 | 570 | ||
GenBank mRNA | AF537131.1 | - | 3 | 524 | ||
AK016223.1 | - | 3 | 535 | |||
AK038879.1 | - | 3 | 325 | |||
AK084539.1 | - | 3 | 570 | |||
AY118229.1 | - | 2 | 262 | |||
BC138370.1 | - | 3 | 295 | 8861390 |