Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Txndc2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001146002.1 | NP_001139474.1 | 3 | 756 | ||
NM_153519.2 | NP_705739.2 | 2 | 584 | |||
GenBank mRNA | AF196282.2 | - | 2 | 584 | ||
BC060981.1 | - | 3 | 721 | 6770744 |