Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: P2rx2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001164833.1 | NP_001158305.1 | 10 | 1094 | ||
NM_001164834.1 | NP_001158306.1 | 10 | 1094 | |||
NM_153400.4 | NP_700449.2 | 10 | 1094 | |||
GenBank mRNA | AB094663.1 | - | 10 | 1014 | ||
AB094664.1 | - | 10 | 1014 | |||
AK141196.1 | - | 10 | 1053 | |||
AY044240.2 | - | 10 | 1270 | |||
BC106963.1 | - | 7 | 1258 | |||
BC106964.1 | - | 10 | 901 | 40039097 |