Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gnl3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153547.6 | NP_705775.2 | 10, 11 | 1227 | ||
GenBank mRNA | AK077523.1 | - | 10, 11 | 1158 | ||
AK167183.1 | - | 10, 11 | 1111 | |||
AY181025.1 | - | 10, 11 | 1085 | |||
AY185498.1 | - | 10, 11 | 1085 | |||
BC018560.1 | - | 10, 11 | 1066 | |||
BC037996.1 | - | 10, 11 | 1087 | 5052858 |