Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Erlin2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153592.2 | NP_705820.1 | 2 | 215 | ||
GenBank mRNA | AK030557.1 | - | 2 | 159 | ||
BC036333.1 | - | 2 | 180 | 3590904 | ||
BC038374.1 | - | 2 | 155 | 5355446 | ||
BY134750.1 | - | 2 | 217 |