Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Flywch1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153791.2 | NP_722486.2 | 2 | 385 | ||
GenBank mRNA | AK168389.1 | - | 2 | 317 | ||
BC038031.1 | - | 1 | 677 | 4021754 | ||
CA321280.1 | - | 1 | 46 | |||
CD553578.1 | - | 2 | 297 |