Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Traf7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001172113.1 | NP_001165584.1 | 6 | 633 | ||
NM_001172114.1 | NP_001165585.1 | 5 | 519 | |||
NM_153792.2 | NP_722487.2 | 6 | 653 | |||
GenBank mRNA | AK088431.1 | - | 5 | 428 | ||
AK131617.1 | - | 6 | 594 | |||
AK185224.1 | - | 1 | 79 | |||
AK185535.1 | - | 1 | 79 | |||
AK189438.1 | - | 1 | 79 | |||
AK216596.1 | - | 1 | 73 | |||
BC008598.1 | - | 6 | 556 | 3499933 |