Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Flywch1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153791.2 | NP_722486.2 | 2 | 467 | ||
GenBank mRNA | AK034869.1 | - | 1 | 61 | ||
AK168389.1 | - | 2 | 399 | |||
BC038031.1 | - | 1 | 759 | 4021754 | ||
CA321280.1 | - | 1 | 128 | |||
CD553578.1 | - | 2 | 379 |