Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Yrdc | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153566.2 | NP_705794.2 | 5 | 885 | ||
GenBank mRNA | AK153627.1 | - | 5 | 885 | ||
AK219737.1 | - | 3 | 347 | |||
BC023823.1 | - | 5 | 871 | 5343817 |