Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tmem121 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153776.2 | NP_722471.2 | 2 | 1564 | ||
GenBank mRNA | AF488729.1 | - | 2 | 1544 | ||
AK010454.2 | - | 2 | 1544 | |||
BC023493.1 | - | 1 | 944 | |||
BC043474.1 | - | 1 | 1521 | 5364389 |