Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gm1976 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK049147.1 | - | 1 | 676 |
Gene Symbol: 2610044O15Rik8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153780.3 | NP_722475.1 | 5 | 853 | ||
GenBank mRNA | AK011776.1 | - | 5 | 830 | ||
AK138138.1 | - | 5 | 854 | |||
BC023890.1 | - | 5 | 812 | 5252244 |
Gene Symbol: Zfp825 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC094048.1 | - | 4 | 1429 | 5251253 |