Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Bclaf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001025392.1 | NP_001020563.1 | 8, 9 | 2295 | ||
NM_001025393.1 | NP_001020564.1 | 8, 9 | 2295 | |||
NM_153787.2 | NP_722482.1 | 8, 9 | 2289 | |||
GenBank mRNA | AK079110.1 | - | 4, 5 | 545 | ||
AK088741.1 | - | 8, 9 | 2230 | |||
AK129071.1 | - | 5, 6 | 1579 | |||
AK141834.1 | - | 8, 9 | 2242 | |||
AK149724.1 | - | 8, 9 | 2248 | |||
AK150017.1 | - | 8, 9 | 2248 | |||
AK161608.1 | - | 1184 | ||||
AK161692.1 | - | 8, 9 | 2293 | |||
BC029682.1 | - | 8, 9 | 2226 | |||
BC034300.1 | - | 8, 9 | 2221 | 4507876 | ||
BC086624.1 | - | 8, 9 | 2277 |