Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Scarf2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153790.2 | NP_722485.1 | 11 | 2978 | ||
GenBank mRNA | AB024432.1 | - | 11 | 2945 | ||
AF522197.1 | - | 11 | 2979 | |||
AK142378.1 | - | 10 | 3197 | |||
AK143208.1 | - | 11 | 2822 | |||
AK170331.1 | - | 5 | 2314 | |||
BC059226.1 | - | 10 | 3115 | |||
BC060084.1 | - | 10 | 3115 |