Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Elof1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_170777.3 | NP_740747.1 | 2 | 711 | ||
GenBank mRNA | AK002505.1 | - | 545 | |||
AK003614.1 | - | 549 | ||||
AK014116.1 | - | 525 | ||||
AK089052.1 | - | 535 | ||||
BC019870.1 | - | 518 | 5123727 | |||
BC024488.1 | - | 530 | 4981209 | |||
BC056225.1 | - | 2 | 711 | 5150539 |