Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Adamts16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172053.2 | NP_742050.2 | 3 | 653 | ||
GenBank mRNA | AK031314.1 | - | 3 | 619 | ||
AK134523.1 | - | 3 | 639 | |||
AK173338.1 | - | 3 | 589 | |||
BC034739.1 | - | 3 | 666 | 4951547 |