Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dio3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172119.2 | NP_742117.2 | 1 | 934 | ||
GenBank mRNA | AK140797.1 | - | 1 | 935 | ||
AK146060.1 | - | 1 | 935 | |||
BC106847.1 | - | 1 | 904 | 40046057 | ||
BC106848.1 | - | 1 | 904 | 40046058 |