Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lpgat1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001134829.1 | NP_001128301.1 | 2 | 413 | ||
NM_172266.3 | NP_758470.1 | 2 | 392 | |||
GenBank mRNA | AK133814.1 | - | 2 | 393 | ||
AK172914.1 | - | 2 | 363 | |||
BC013667.1 | - | 2 | 301 | 4020003 | ||
BY731977.1 | - | 2 | 413 |