Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: B3gat2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172124.2 | NP_742122.2 | 2 | 1433 | ||
GenBank mRNA | AB055902.1 | - | 2 | 1379 | ||
AK048146.1 | - | 2 | 1349 | |||
AK052640.1 | - | 2 | 1433 | |||
AK136293.1 | - | 2 | 354 | |||
AK220433.1 | - | 1341 | ||||
BC056368.1 | - | 2 | 1325 | 6827380 | ||
BC058082.1 | - | 2 | 1325 | 6827380 | ||
BC092280.1 | - | 3 | 1098 |