Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Phyhd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001252568.2 | NP_001239497.1 | 3 | 581 | ||
NM_001252570.1 | NP_001239499.1 | 2 | 202 | |||
NM_001252571.1 | NP_001239500.1 | 2 | 202 | |||
NM_001281829.1 | NP_001268758.1 | 3 | 228 | |||
NM_172267.4 | NP_758471.1 | 3 | 581 | |||
GenBank mRNA | AI182929.1 | - | 3 | 203 | ||
AK005293.1 | - | 3 | 582 | |||
AK046118.1 | - | 2 | 171 | |||
AK138607.1 | - | 3 | 256 | |||
BC039982.1 | - | 3 | 223 | |||
BC132271.1 | - | 3 | 192 | 40130548 | ||
BC132273.1 | - | 3 | 192 | 40130550 | ||
BC145356.1 | - | 3 | 192 | 9053914 | ||
BC171925.1 | - | 3 | 192 | |||
BQ829083.1 | - | 2 | 198 | |||
BY196857.1 | - | 3 | 230 | |||
BY344206.1 | - | 3 | 235 | |||
CX235839.1 | - | 1 | 48 |