Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Megf6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001162977.1 | NP_001156449.1 | 32 | 4351 | ||
GenBank mRNA | AK134303.1 | - | 19 | 2345 | ||
AK173032.1 | - | 15 | 1830 | |||
AK196435.1 | - | 1 | 20 | |||
BC031402.1 | - | 12 | 1389 | |||
BC039980.1 | - | 12 | 1400 | |||
BC058571.1 | - | 11 | 1293 |