Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Jade1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001130184.1 | NP_001123656.1 | 4, 5 | 419 | ||
NM_001130185.1 | NP_001123657.1 | 4, 5 | 499 | |||
NM_001130186.1 | NP_001123658.1 | 4, 5 | 457 | |||
NM_172303.4 | NP_758507.3 | 4, 5 | 517 | |||
GenBank mRNA | AK049332.1 | - | 4, 5 | 422 | ||
AK129445.1 | - | 4, 5 | 412 | |||
AK147466.1 | - | 4, 5 | 379 | |||
BC020316.2 | - | 4, 5 | 439 | 5250231 | ||
BY060282.1 | - | 4, 5 | 415 |