Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dnajc16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172338.2 | NP_758841.1 | 15 | 3878 | ||
GenBank mRNA | AK051048.1 | - | 15 | 3880 | ||
AK122407.1 | - | 4115 | ||||
AK154946.1 | - | 3899 | ||||
AK164023.1 | - | 15 | 3894 | |||
BC046424.1 | - | 9 | 2873 | 3989412 |