Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SHBG | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001040.4 | NP_001031.2 | 5 | 783 | ||
NM_001146279.2 | NP_001139751.1 | 5 | 729 | |||
NM_001146280.2 | NP_001139752.1 | 5 | 783 | |||
NM_001146281.2 | NP_001139753.1 | 5 | 783 | |||
NM_001289113.1 | NP_001276042.1 | 5 | 776 | |||
NM_001289114.1 | NP_001276043.1 | 5 | 639 | |||
NM_001289115.1 | NP_001276044.1 | 5 | 776 | |||
NM_001289116.1 | NP_001276045.1 | 5 | 691 | |||
GenBank mRNA | AK302603.1 | - | 5 | 776 | ||
BC069597.1 | - | 5 | 701 | |||
BC101785.1 | - | 5 | 735 | 8069291 | ||
BC112186.1 | - | 5 | 735 | 8327654 | ||
CD013955.1 | - | 5 | 725 | |||
CD013957.1 | - | 5 | 671 | |||
CD013958.1 | - | 5 | 633 | |||
EU352661.1 | - | 5 | 623 | |||
EU352662.1 | - | 4 | 461 | |||
EU352663.1 | - | 4 | 461 | |||
EU352665.1 | - | 5 | 644 | |||
EU352666.1 | - | 5 | 644 | |||
EU352667.1 | - | 4 | 482 | |||
EU352668.1 | - | 4 | 482 | |||
EU352670.1 | - | 5 | 639 | |||
EU352671.1 | - | 5 | 639 | |||
EU352672.1 | - | 5 | 639 | |||
EU352673.1 | - | 4 | 477 | |||
EU352675.1 | - | 4 | 477 | |||
EU352676.1 | - | 4 | 477 | |||
X05403.1 | - | 5 | 641 | |||
X05792.1 | - | 3 | 346 | |||
X05885.1 | - | 4 | 568 | |||
X16351.1 | - | 5 | 689 |