Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Arhgef10l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001112722.1 | NP_001106193.1 | 6 | 579 | ||
NM_001112723.1 | NP_001106194.1 | 6 | 552 | |||
NM_001290803.1 | NP_001277732.1 | 6 | 561 | |||
NM_172415.3 | NP_766003.3 | 6 | 552 | |||
GenBank mRNA | AK028648.1 | - | 6 | 562 | ||
AK129408.1 | - | 6 | 552 | |||
BB612864.1 | - | 6 | 562 | |||
BC059220.1 | - | 6 | 546 | 6417641 | ||
CF532111.1 | - | 3 | 176 |