Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Celf3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001289613.1 | NP_001276542.1 | 1, 2 | 710 | ||
NM_001289616.1 | NP_001276545.1 | 2, 3 | 195 | |||
NM_001289620.1 | NP_001276549.1 | 2, 3 | 195 | |||
NM_172434.3 | NP_766022.1 | 1, 2 | 710 | |||
GenBank mRNA | AK044069.1 | - | 1, 2 | 143 | ||
AK044506.1 | - | 2, 3 | 159 | |||
AK087536.1 | - | 2, 3 | 155 | |||
AY165052.1 | - | 1, 2 | 428 | |||
BC057553.1 | - | 1, 2 | 765 | 6401356 | ||
BY089288.1 | - | 2, 3 | 196 |